Gene Transcripts
- Gene Symbol
- AP1G1
- Entrez Gene ID
- 164
- Gene Name
- adaptor related protein complex 1 gamma 1 subunit
- Gene Aliases
- ADTG, CLAPG1
- Chromosome Location
- Chr. 16: 71729000 - 71809073 on Build GRCh38
- UniGene ID
- Hs.461253
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001030007.1 |
NP_001025178.1
|
8, 9 |
1069 |
|
NM_001128.5 |
NP_001119.3
|
7, 8 |
1060 |
|
GenBank mRNA |
AB015317.1 |
- |
7, 8 |
765 |
|
AK128078.1 |
- |
10, 11 |
1615 |
|
AK225906.1 |
- |
8, 9 |
828 |
|
AK293233.1 |
- |
8, 9 |
719 |
|
AK294546.1 |
- |
8, 9 |
913 |
|
AK299630.1 |
- |
9, 10 |
1003 |
|
AK314484.1 |
- |
7, 8 |
1065 |
|
BC036283.1 |
- |
8, 9 |
1034 |
4158263 |
Y12226.1 |
- |
8, 9 |
783 |
|
View Full Product Details