Gene Transcripts

Gene Symbol
Trim12c
Entrez Gene ID
319236
Gene Name
tripartite motif-containing 12C
Gene Aliases
9230105E10Rik, Trim12-2
Chromosome Location
Chr. 7: 104338754 - 104353358 on Build GRCm38
UniGene ID
Mm.425502
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146007.1 NP_001139479.1 4 799
NM_175677.4 NP_783608.3 4 680
GenBank mRNA AK033764.1 - 4 800
AK050485.1 - 4 804
AK146437.1 - 4 759
BC038937.1 - 4 680 2648191

View Full Product Details