Gene Transcripts
- Gene Symbol
- Wdr59
- Entrez Gene ID
- 319481
- Gene Name
- WD repeat domain 59
- Gene Aliases
- 5430401O09Rik, Cdw12
- Chromosome Location
- Chr. 8: 111448781 - 111522107 on Build GRCm38
- UniGene ID
- Mm.247294
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001170742.1 |
NP_001164213.1
|
19, 20 |
2121 |
|
NM_001170743.1 |
NP_001164214.1
|
20, 21 |
2178 |
|
NM_176923.4 |
NP_795897.2
|
19, 20 |
2121 |
|
GenBank mRNA |
AK030644.1 |
- |
19, 20 |
2002 |
|
AK040807.1 |
- |
4, 5 |
408 |
|
AK136303.1 |
- |
15, 16 |
1673 |
|
AK170910.1 |
- |
19, 20 |
2120 |
|
AK173301.1 |
- |
20, 21 |
2141 |
|
BC066082.1 |
- |
19, 20 |
2106 |
30355918 |
EF011622.1 |
- |
19, 20 |
2106 |
|
View Full Product Details