Gene Transcripts

Gene Symbol
Auts2
Entrez Gene ID
319974
Gene Name
autism susceptibility candidate 2
Gene Aliases
2700063G02Rik, A730011F23Rik, D830032G16Rik
Chromosome Location
Chr. 5: 131437682 - 132542343 on Build GRCm38
UniGene ID
Mm.409595
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177047.3 NP_796021.2 1, 2 836
GenBank mRNA AK052418.1 - 1, 2 898
AK077988.1 - 1, 2 1293
AK147478.1 - 1835
BC096021.1 - 1, 2 835
BC151031.1 - 1, 2 850
BC151047.1 - 1, 2 849

View Full Product Details