Gene Transcripts

Gene Symbol
Alg6
Entrez Gene ID
320438
Gene Name
asparagine-linked glycosylation 6 (alpha-1,3,-glucosyltransferase)
Gene Aliases
E230028F23Rik
Chromosome Location
Chr. 4: 99715599 - 99763460 on Build GRCm38
UniGene ID
Mm.32408
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081264.1 NP_001074733.1 3 349
GenBank mRNA AK036792.1 - 3 311
AK087075.1 - 3 316
AK087630.1 - 3 308
AK157903.1 - 3 311
AK171887.1 - 3 302
BC050854.1 - 3 314 6332345
BC080867.1 - 3 289

View Full Product Details