Gene Transcripts

Gene Symbol
Mysm1
Entrez Gene ID
320713
Gene Name
myb-like, SWIRM and MPN domains 1
Gene Aliases
C130067A03Rik, C530050H10Rik
Chromosome Location
Chr. 4: 94942037 - 94979302 on Build GRCm38
UniGene ID
Mm.208868
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177239.2 NP_796213.2 18 2251
GenBank mRNA AK173300.1 - 15 1993
BC150946.1 - 18 2247 9087857
BC151172.1 - 18 2247 9088061

View Full Product Details