Gene Transcripts

Gene Symbol
Med13
Entrez Gene ID
327987
Gene Name
mediator complex subunit 13
Gene Aliases
1110067M05Rik, D030023K18, Thrap1, Trap240
Chromosome Location
Chr. 11: 86265715 - 86357525 on Build GRCm38
UniGene ID
Mm.341886
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080931.1 NP_001074400.1 23 5260
GenBank mRNA AK037303.1 - 9 2470
AK129168.1 - 6 1367

View Full Product Details