Gene Transcripts

Gene Symbol
Spata21
Entrez Gene ID
329972
Gene Name
spermatogenesis associated 21
Gene Aliases
4933414G08
Chromosome Location
Chr. 4: 141088322 - 141112760 on Build GRCm38
UniGene ID
Mm.247201
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177867.3 NP_808535.1 10 1675
GenBank mRNA AK077155.1 - 10 1676
BC112427.1 - 9 1384 40062127
BC113132.1 - 9 1384 40062125

View Full Product Details