Gene Transcripts

Gene Symbol
B4galnt4
Entrez Gene ID
330671
Gene Name
beta-1,4-N-acetyl-galactosaminyl transferase 4
Gene Aliases
BC038881, Gm1095, beta4GalNAcT4
Chromosome Location
Chr. 7: 141061109 - 141072119 on Build GRCm38
UniGene ID
Mm.133741
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177897.3 NP_808565.2 16, 17 2631
GenBank mRNA AB114827.1 - 16, 17 2631
BC038881.1 - 2, 3 578
BC062653.1 - 12, 13 2321
BC079533.1 - 5, 6 1534

View Full Product Details