Gene Transcripts

Gene Symbol
Plscr5
Entrez Gene ID
100504689
Gene Name
phospholipid scramblase family, member 5
Gene Aliases
331000
Chromosome Location
Chr. 9: 92186511 - 92212539 on Build GRCm38
UniGene ID
Mm.464262
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195693.1 NP_001182622.1 2 362

View Full Product Details