Gene Transcripts

Gene Symbol
Gsdmc2
Entrez Gene ID
331063
Gene Name
gasdermin C2
Gene Aliases
9130023H10, AI987692
Chromosome Location
Chr. 15: 63824328 - 63845196 on Build GRCm38
UniGene ID
Mm.273185
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168274.1 NP_001161746.1 1729
NM_177912.4 NP_808580.2 1494
GenBank mRNA AK033603.1 - 1495
BC113155.1 - 1715 40058514

View Full Product Details