Gene Transcripts

Gene Symbol
Dgkk
Entrez Gene ID
331374
Gene Name
diacylglycerol kinase kappa
Gene Aliases
Gm360
Chromosome Location
Chr. X: 6779306 - 6779636 on Build GRCm38
UniGene ID
Mm.392752
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177914.3 NP_808582.3 2294
GenBank mRNA BC075627.1 - 2294

View Full Product Details