Gene Transcripts

Gene Symbol
Bend3
Entrez Gene ID
331623
Gene Name
BEN domain containing 3
Gene Aliases
mKIAA1553
Chromosome Location
Chr. 10: 43479140 - 43515417 on Build GRCm38
UniGene ID
Mm.490388
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199028.2 NP_950193.1 4 2247
GenBank mRNA AK122525.1 - 2 1853
BC060240.1 - 4 2247 6856073

View Full Product Details