Gene Transcripts

Gene Symbol
Krt78
Entrez Gene ID
332131
Gene Name
keratin 78
Gene Aliases
2310030B04Rik, Kb40
Chromosome Location
Chr. 15: 101946001 - 101996609 on Build GRCm38
UniGene ID
Mm.325105
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_212487.4 NP_997652.4 7 1294
GenBank mRNA AK146611.1 - 7 1291
AY651827.1 - 7 1119
BC119600.1 - 7 1231
BC119601.1 - 7 1231
BC127241.1 - 7 1231
BC127618.1 - 7 1231

View Full Product Details