Gene Transcripts

Gene Symbol
Mrs2
Entrez Gene ID
380836
Gene Name
MRS2 magnesium homeostasis factor homolog (S. cerevisiae)
Gene Aliases
Gm902, HPT, Mrs2l, RPT
Chromosome Location
Chr. 13: 24987483 - 25020358 on Build GRCm38
UniGene ID
Mm.387097
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013389.2 NP_001013407.2 10, 11 1274
GenBank mRNA AK136100.1 - 5, 6 563
BB015377.2 - 3, 4 375

View Full Product Details