Gene Transcripts

Gene Symbol
Parl
Entrez Gene ID
381038
Gene Name
presenilin associated, rhomboid-like
Gene Aliases
D16Ertd607e, PRO2207, PSARL1, PSENIP2, Psarl
Chromosome Location
Chr. 16: 20279820 - 20302395 on Build GRCm38
UniGene ID
Mm.371737
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005767.4 NP_001005767.1 2 214
GenBank mRNA AK039142.1 - 2 193
AK150980.1 - 2 222
AK151652.1 - 2 213
AK179606.1 - 2 79
AK181973.1 - 2 80
AK207637.1 - 2 223
AK216516.1 - 2 79
BC083153.1 - 2 214 30728006

View Full Product Details