Gene Transcripts

Gene Symbol
Cdkl5
Entrez Gene ID
382253
Gene Name
cyclin-dependent kinase-like 5
Gene Aliases
BC038161, Stk9
Chromosome Location
Chr. X: 160784308 - 160994681 on Build GRCm38
UniGene ID
Mm.443717
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024624.2 NP_001019795.1 18 2769
GenBank mRNA AK139220.1 - 18 2741
BC038161.1 - 4 327

View Full Product Details