Gene Transcripts

Gene Symbol
Gps2
Entrez Gene ID
56310
Gene Name
G protein pathway suppressor 2
Gene Aliases
AI505953
Chromosome Location
Chr. 11: 69913980 - 69917150 on Build GRCm38
UniGene ID
Mm.477738
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019726.3 NP_062700.2 6 642
GenBank mRNA BC011317.1 - 6 642
BC138879.1 - 6 598 8861901
BC138880.1 - 6 599 8861902

View Full Product Details