Gene Transcripts

Gene Symbol
Trim56
Entrez Gene ID
384309
Gene Name
tripartite motif-containing 56
Gene Aliases
A130009K11Rik, Gm452, RNF109
Chromosome Location
Chr. 5: 137111286 - 137116466 on Build GRCm38
UniGene ID
Mm.100647
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201373.4 NP_958761.1 3 317
GenBank mRNA AK037350.1 - 3 318
BC045615.1 - 3 307 4976515

View Full Product Details