Gene Transcripts
- Gene Symbol
- Trav7d-4
- Entrez Gene ID
- 382180
- Gene Name
- T cell receptor alpha variable 7D-4
- Gene Aliases
- Gm13959, OTTMUSG00000015181
- Chromosome Location
- Chr. 14: 52769863 - 52770392 on Build GRCm38
- UniGene ID
- Mm.429462
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY639036.1 |
- |
2 |
136 |
|
- Gene Symbol
- Trav7-4
- Entrez Gene ID
- 386450
- Gene Name
- T cell receptor alpha variable 7-4
- Gene Aliases
- Gm13949, OTTMUSG00000015161
- Chromosome Location
- Chr. 14: 53461209 - 53461738 on Build GRCm38
- UniGene ID
- Mm.425889
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
X56720.1 |
- |
|
125 |
|
X56721.1 |
- |
|
125 |
|
X63928.1 |
- |
|
136 |
|
View Full Product Details