Gene Transcripts
- Gene Symbol
- Tas2r130
- Entrez Gene ID
- 387355
- Gene Name
- taste receptor, type 2, member 130
- Gene Aliases
- STC74, T2R30, Tas2r30, Tas2r7, mt2r42
- Chromosome Location
- Chr. 6: 131629892 - 131630830 on Build GRCm38
- UniGene ID
- Mm.377916
- Species
- Mus musculus
- Gene Symbol
- Stc7-1
- Entrez Gene ID
- 100500796
- Gene Name
- T2R family member
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF412302.1 |
- |
1 |
512 |
|
View Full Product Details