Gene Transcripts

Gene Symbol
Arpc2
Entrez Gene ID
76709
Gene Name
actin related protein 2/3 complex, subunit 2
Gene Aliases
2210023N03Rik, 34kDa, p34-Arc
Chromosome Location
Chr. 1: 74236504 - 74268213 on Build GRCm38
UniGene ID
Mm.337038
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029711.1 NP_083987.1 4, 5 546
GenBank mRNA AK083263.1 - 4, 5 356
AK151513.1 - 4, 5 520
AK152356.1 - 5, 6 371
AK170061.1 - 5, 6 345
BC099504.1 - 3, 4 198
BC115750.1 - 4, 5 344 40051600

View Full Product Details