Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025526.4 | NP_079802.1 | 2 | 180 | |
GenBank mRNA | AK009021.1 | - | 2 | 181 | |
AK009421.1 | - | 2 | 184 | ||
AK039834.1 | - | 2 | 222 | ||
AK083247.1 | - | 2 | 188 | ||
AK157864.1 | - | 2 | 189 | ||
BC028800.1 | - | 2 | 158 | ||
BC048409.1 | - | 2 | 177 | 6484953 | |
BC096049.1 | - | 2 | 178 | 3466271 |