Gene Transcripts

Gene Symbol
Spin2d
Entrez Gene ID
100504429
Gene Name
spindlin family, member 2D
Gene Aliases
4930408F14Rik
Chromosome Location
Chr. X: 73175302 - 73176989 on Build GRCm38
UniGene ID
Mm.307966
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243002.1 NP_001229931.1 2 437
GenBank mRNA AK015110.1 - 2 438

Gene Symbol
Gm21637
Entrez Gene ID
100862314
Gene Name
predicted gene, 21637
Gene Aliases
Chromosome Location
Chr. X: 31960926 - 31962821 on Build GRCm38
UniGene ID
Mm.431329
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270685.1 NP_001257614.1 1 228
GenBank mRNA BC117726.1 - 1 228

Gene Symbol
Spin2-ps1
Entrez Gene ID
74851
Gene Name
spindlin family, member 2, pseudogene 1
Gene Aliases
Gm14354, OTTMUSG00000016414
Chromosome Location
Chr. X: 3835879 - 3836656 on Build GRCm38
UniGene ID
Mm.431329
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029106.2 NP_083382.2 1 228

View Full Product Details