Gene Transcripts

Gene Symbol
Cers3
Entrez Gene ID
545975
Gene Name
ceramide synthase 3
Gene Aliases
4930550L11Rik, Gm488, Lass3, T3L
Chromosome Location
Chr. 7: 66733178 - 66823697 on Build GRCm38
UniGene ID
Mm.380104
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164201.1 NP_001157673.1 8 952
GenBank mRNA DQ358087.1 - 7 710
DQ646881.1 - 8 831

View Full Product Details