Gene Transcripts

Gene Symbol
Card6
Entrez Gene ID
239319
Gene Name
caspase recruitment domain family, member 6
Gene Aliases
D730008L15
Chromosome Location
Chr. 15: 5096920 - 5116483 on Build GRCm38
UniGene ID
Mm.31252
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163138.1 NP_001156610.1 4 1017
GenBank mRNA AK052796.1 - 3 907

View Full Product Details