Gene Transcripts

Gene Symbol
Xlr3b
Entrez Gene ID
574437
Gene Name
X-linked lymphocyte-regulated 3B
Gene Aliases
EG574437, Xlr3
Chromosome Location
Chr. X: 73192152 - 73202930 on Build GRCm38
UniGene ID
Mm.482923
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081643.1 NP_001075112.1 6, 7 528
GenBank mRNA AK146398.1 - 5, 6 1297
BC038862.1 - 6, 7 491
BC057003.1 - 4, 5 2691
BC064787.1 - 6, 7 485 30046437

Gene Symbol
Xlr3a
Entrez Gene ID
22445
Gene Name
X-linked lymphocyte-regulated 3A
Gene Aliases
Xlr3
Chromosome Location
Chr. X: 73086293 - 73097095 on Build GRCm38
UniGene ID
Mm.475098
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110784.1 NP_001104254.1 6, 7 557
GenBank mRNA BC003812.1 - 6, 7 503
BC093495.1 - 480 3593889
BC107255.1 - 466 40052405
BC107283.1 - 467 40052875
BC127145.1 - 467 40052876
BC127146.1 - 468 40052877
BQ885410.1 - 6, 7 497
DV662513.1 - 6, 7 505
U02599.1 - 5, 6 450

Gene Symbol
Xlr3c
Entrez Gene ID
22446
Gene Name
X-linked lymphocyte-regulated 3C
Gene Aliases
Xlr3b
Chromosome Location
Chr. X: 73254538 - 73265571 on Build GRCm38
UniGene ID
Mm.336117
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011727.2 NP_035857.2 6, 7 497
GenBank mRNA L22977.1 - 482
U02600.1 - 6, 7 497

View Full Product Details