Gene Transcripts
- Gene Symbol
- RPL9
- Entrez Gene ID
- 6133
- Gene Name
- ribosomal protein L9
- Gene Aliases
- L9, NPC-A-16
- Chromosome Location
- Chr. 4: 39454124 - 39458948 on Build GRCh38
- UniGene ID
- Hs.412370
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000661.4 |
NP_000652.2
|
2, 3 |
104 |
|
NM_001024921.3 |
NP_001020092.1
|
2, 3 |
262 |
|
GenBank mRNA |
AB062431.1 |
- |
1 |
51 |
|
AK130966.1 |
- |
1 |
67 |
|
AK297178.1 |
- |
2, 3 |
87 |
|
AK303908.1 |
- |
1 |
67 |
|
AK312241.1 |
- |
1 |
71 |
|
AL039355.2 |
- |
2, 3 |
237 |
|
AY320412.1 |
- |
2, 3 |
87 |
|
AY376242.1 |
- |
1, 2 |
221 |
|
AY597415.1 |
- |
1 |
78 |
|
BC000483.2 |
- |
1 |
56 |
2964733 |
BC004156.2 |
- |
1 |
50 |
2961026 |
BC004206.2 |
- |
1 |
50 |
2961026 |
BC007967.2 |
- |
1 |
106 |
4304670 |
BC012149.1 |
- |
1 |
106 |
4554838 |
BC031906.1 |
- |
1, 2 |
51 |
4996986 |
BC066318.1 |
- |
1 |
71 |
5277491 |
BC070214.1 |
- |
1 |
110 |
6605055 |
BC100285.1 |
- |
1, 2 |
211 |
6022858 |
BG829769.1 |
- |
1, 2 |
52 |
|
BM841336.1 |
- |
2, 3 |
104 |
|
D14531.1 |
- |
1, 2 |
51 |
|
D16961.1 |
- |
1 |
24 |
|
U09953.1 |
- |
2, 3 |
74 |
|
U21138.1 |
- |
1 |
51 |
|
View Full Product Details