Gene Transcripts

Gene Symbol
Lekr1
Entrez Gene ID
624866
Gene Name
leucine, glutamate and lysine rich 1
Gene Aliases
EG546798, EG624866, Gm6534
Chromosome Location
Chr. 3: 65659539 - 65832480 on Build GRCm38
UniGene ID
Mm.296317
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037923.4 NP_001033012.2 3 219
NM_001166659.1 NP_001160131.1 3 219
GenBank mRNA AK137452.1 - 3 179
BC147278.1 - 2 148 9056357
BC147279.1 - 2 148 9056330
CJ055787.1 - 3 214
CJ137734.1 - 3 220

View Full Product Details