Gene Transcripts

Gene Symbol
Vmn2r83
Entrez Gene ID
625029
Gene Name
vomeronasal 2, receptor 83
Gene Aliases
EG625029
Chromosome Location
Chr. 10: 79468958 - 79492154 on Build GRCm38
UniGene ID
Mm.469865
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104537.1 NP_001098007.1 3 952

View Full Product Details