Gene Transcripts

Gene Symbol
Vmn2r86
Entrez Gene ID
625109
Gene Name
vomeronasal 2, receptor 86
Gene Aliases
EG625109
Chromosome Location
Chr. 10: 130446199 - 130455894 on Build GRCm38
UniGene ID
Mm.469845
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001103365.1 NP_001096835.1 4 1466

View Full Product Details