Gene Transcripts

Gene Symbol
Rybp
Entrez Gene ID
56353
Gene Name
RING1 and YY1 binding protein
Gene Aliases
2410018J24Rik, DEDAF, YEAF1
Chromosome Location
Chr. 6: 100228565 - 100287358 on Build GRCm38
UniGene ID
Mm.321633
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019743.3 NP_062717.2 4 466
GenBank mRNA AF101779.1 - 4 523
AK033024.1 - 4 505
BC053016.1 - 4 466 6406584
BC080287.1 - 4 549 6410301
BC138324.1 - 4 448 8861344
BC138325.1 - 4 448 8861345

View Full Product Details