Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007917.3 | NP_031943.3 | 5 | 373 | |
GenBank mRNA | AK082867.1 | - | 4 | 254 | |
AK146757.1 | - | 4 | 253 | ||
AK167097.1 | - | 4 | 262 | ||
BC010759.1 | - | 4 | 247 | 4216609 | |
BC085087.1 | - | 4 | 245 | 6438419 | |
CN714483.1 | - | 5 | 373 | ||
M61731.1 | - | 4 | 259 |