Gene Transcripts

Gene Symbol
Ssxb8
Entrez Gene ID
631002
Gene Name
synovial sarcoma, X member B, breakpoint 8
Gene Aliases
Chromosome Location
Chr. X: 8684843 - 8690883 on Build GRCm38
UniGene ID
Mm.379565
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081565.3 NP_001075034.3 5 383
GenBank mRNA AY244799.1 - 384

View Full Product Details