Gene Transcripts

Gene Symbol
RPS21
Entrez Gene ID
6227
Gene Name
ribosomal protein S21
Gene Aliases
HLDF, S21
Chromosome Location
Chr. 20: 62387065 - 62388520 on Build GRCh38
UniGene ID
Hs.190968
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024.3 NP_001015.1 4 244
GenBank mRNA AV758927.1 - 205
BC018140.1 - 3 181 3903320
BC021990.1 - 4 209 4066330
BM840443.1 - 4 244
CF124568.1 - 4 285
CR542132.1 - 3 154
L04483.1 - 4 192
X79563.2 - 4 190

Gene Symbol
TAPT1-AS1
Entrez Gene ID
202020
Gene Name
TAPT1 antisense RNA 1 (head to head)
Gene Aliases
Chromosome Location
Chr. 4: 16226663 - 16258187 on Build GRCh38
UniGene ID
Hs.445315
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AI770046.1 - 1 576

View Full Product Details