Gene Transcripts

Gene Symbol
Tsr1
Entrez Gene ID
104662
Gene Name
TSR1 20S rRNA accumulation
Gene Aliases
AU040765, AW550801, mKIAA1401
Chromosome Location
Chr. 11: 74898080 - 74909340 on Build GRCm38
UniGene ID
Mm.220843
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177325.3 NP_796299.2 10 1741
GenBank mRNA AK129350.1 - 10 1695
AK154063.1 - 10 1728
AK162686.1 - 4 527
BC031531.1 - 10 1714 4014819

View Full Product Details