Gene Transcripts

Gene Symbol
Clec2l
Entrez Gene ID
665180
Gene Name
C-type lectin domain family 2, member L
Gene Aliases
AI414054, AW122724, Gm1064
Chromosome Location
Chr. 6: 38663069 - 38680865 on Build GRCm38
UniGene ID
Mm.349066
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101507.1 NP_001094977.1 5 1017
GenBank mRNA AI627165.1 - 1 259

View Full Product Details