Gene Transcripts

Gene Symbol
SELPLG
Entrez Gene ID
6404
Gene Name
selectin P ligand
Gene Aliases
CD162, CLA, PSGL-1, PSGL1
Chromosome Location
Chr. 12: 108621895 - 108633894 on Build GRCh38
UniGene ID
Hs.591014
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001206609.1 NP_001193538.1 2 1135
NM_003006.4 NP_002997.2 2 1070
GenBank mRNA AK098315.1 - 4 989
AK127638.1 - 3 1021
AK225644.1 - 2 966
AK290395.1 - 3 1007
AK295238.1 - 3 852
AK298738.1 - 2 1068
AK298742.1 - 2 1107
AK300057.1 - 3 962
BC029782.1 - 3 1019 5180824
U02297.1 - 3 939

View Full Product Details