Gene Transcripts

Gene Symbol
Rhox5
Entrez Gene ID
24631
Gene Name
reproductive homeobox 5
Gene Aliases
Pem
Chromosome Location
Chr. X: 123900363 - 123916988 on Build Rnor_6.0
UniGene ID
Rn.44483
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022175.1 NP_071511.1 1, 2 92
GenBank mRNA DQ058653.1 - 1, 2 92

View Full Product Details