Gene Transcripts

Gene Symbol
SLC2A4
Entrez Gene ID
6517
Gene Name
solute carrier family 2 member 4
Gene Aliases
GLUT4
Chromosome Location
Chr. 17: 7281735 - 7288048 on Build GRCh38
UniGene ID
Hs.380691
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042.2 NP_001033.1 2 316
GenBank mRNA AK300899.1 - 2 383
AK313372.1 - 2 330
AL521328.3 - 2 315
BC034387.1 - 2 316 5187454
BC069615.1 - 2 211 7262457
BC069621.1 - 2 211 7262469
BC113592.1 - 2 143 8322644
BC126164.1 - 2 143 8991880
M20747.1 - 2 261

View Full Product Details