Gene Transcripts

Gene Symbol
SNRPA1
Entrez Gene ID
6627
Gene Name
small nuclear ribonucleoprotein polypeptide A'
Gene Aliases
Lea1
Chromosome Location
Chr. 15: 101281510 - 101295282 on Build GRCh38
UniGene ID
Hs.528763
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003090.3 NP_003081.2 2 224
NR_135508.1 2 224
GenBank mRNA AK223070.1 - 2 181
AK290078.1 - 2 180
AK312200.1 - 2 155
AL538391.3 - 2 223
BC022816.1 - 2 177 4904205
BC067846.1 - 2 193 30330402
BC071717.1 - 1 132 4514975
X13482.1 - 2 176

View Full Product Details