Gene Transcripts

Gene Symbol
Ly6c
Entrez Gene ID
56778
Gene Name
Ly6-C antigen
Gene Aliases
Chromosome Location
Chr. 7|NW_007906224: 20092 - 23957 on Build Rnor_6.0
UniGene ID
Rn.40119
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020103.1 NP_064488.1 2, 3 186
XM_006257716.2 XP_006257778.1 3, 4 291
GenBank mRNA FQ230231.1 - 3, 4 244
FQ230335.1 - 3, 4 245
FQ232627.1 - 3, 4 245
FQ234379.1 - 3, 4 244
FQ235068.1 - 3, 4 245
M30691.1 - 2, 3 184

Gene Symbol
LOC100911104
Entrez Gene ID
100911104
Gene Name
lymphocyte antigen 6B-like
Gene Aliases
Chromosome Location
Chr. 14: 22868518 - 22872383 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003751343.2 XP_003751391.1 3, 4 291

View Full Product Details