Gene Transcripts

Gene Symbol
Nckap1
Entrez Gene ID
58823
Gene Name
NCK-associated protein 1
Gene Aliases
Chromosome Location
Chr. 3: 67725180 - 67804680 on Build Rnor_6.0
UniGene ID
Rn.2611
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031618.1 NP_113806.1 2644
GenBank mRNA BC092610.1 - 5 597
D84346.1 - 22 2548

View Full Product Details