Gene Transcripts

Gene Symbol
Cabs1
Entrez Gene ID
64029
Gene Name
calcium-binding protein, spermatid-specific 1
Gene Aliases
Clph, Rsd6
Chromosome Location
Chr. 14: 21746294 - 21748356 on Build Rnor_6.0
UniGene ID
Rn.54001
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022263.2 NP_071599.2 1 720
GenBank mRNA AF271155.1 - 1 416
BC079062.1 - 1 720 7115934

View Full Product Details