Gene Transcripts

Gene Symbol
UQCR11
Entrez Gene ID
10975
Gene Name
ubiquinol-cytochrome c reductase, complex III subunit XI
Gene Aliases
0710008D09Rik, QCR10, UQCR
Chromosome Location
Chr. 19: 1597155 - 1605484 on Build GRCh38
UniGene ID
Hs.8372
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006830.3 NP_006821.1 3 1138
GenBank mRNA BC000462.2 - 3 1086
BC003594.1 - 3 1086 2822699

View Full Product Details