Gene Transcripts

Gene Symbol
Isl1
Entrez Gene ID
64444
Gene Name
ISL LIM homeobox 1
Gene Aliases
Isl-1, isl-1=homeobox
Chromosome Location
Chr. 2: 48488911 - 48501217 on Build Rnor_6.0
UniGene ID
Rn.36202
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017339.3 NP_059035.3 5 855
GenBank mRNA AY557632.1 - 5 781
S69329.1 - 5 855
X53258.1 - 5 958

View Full Product Details