Gene Transcripts
- Gene Symbol
- Golga2
- Entrez Gene ID
- 64528
- Gene Name
- golgin A2
- Gene Aliases
- Gm130
- Chromosome Location
- Chr. 3: 11317328 - 11337569 on Build Rnor_6.0
- UniGene ID
- Rn.6165
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022596.2 |
NP_072118.2
|
7, 8 |
589 |
|
XM_006233992.2 |
XP_006234054.1
|
8, 9 |
639 |
|
XM_006233993.2 |
XP_006234055.1
|
8, 9 |
639 |
|
XM_006233994.2 |
XP_006234056.1
|
8, 9 |
639 |
|
XM_006233995.2 |
XP_006234057.1
|
7, 8 |
558 |
|
XM_006233997.2 |
XP_006234059.1
|
7, 8 |
558 |
|
XM_006233998.2 |
XP_006234060.1
|
7, 8 |
558 |
|
XM_008761774.1 |
XP_008759996.1
|
6, 7 |
463 |
|
GenBank mRNA |
FN805927.1 |
- |
7, 8 |
482 |
|
FQ147221.1 |
- |
1, 2 |
23 |
|
U35022.2 |
- |
7, 8 |
589 |
|
View Full Product Details