Gene Transcripts

Gene Symbol
Vdac2
Entrez Gene ID
83531
Gene Name
voltage-dependent anion channel 2
Gene Aliases
Chromosome Location
Chr. 15: 2634622 - 2648548 on Build Rnor_6.0
UniGene ID
Rn.162037
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031354.1 NP_112644.1 7 786
XM_006251661.1 XP_006251723.1 8 907
GenBank mRNA AB039663.1 - 7 786
AF268468.1 - 6 622
BC063164.1 - 7 755 6921675
FQ214165.1 - 4 511
FQ217146.1 - 4 511
FQ219952.1 - 4 511
FQ228796.1 - 4 511

Gene Symbol
LOC362126
Entrez Gene ID
362126
Gene Name
similar to Voltage-dependent anion-selective channel protein 2 (VDAC-2) (mVDAC2) (mVDAC6) (Outer mitochondrial membrane protein porin 2)
Gene Aliases
Chromosome Location
Chr. 3: 23762540 - 23769642 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008761811.1 XP_008760033.1 1 615

View Full Product Details