Gene Transcripts

Gene Symbol
Atp6ap1
Entrez Gene ID
83615
Gene Name
ATPase, H+ transporting, lysosomal accessory protein 1
Gene Aliases
Atp6s1, C7-1
Chromosome Location
Chr. X: 156407973 - 156415053 on Build Rnor_6.0
UniGene ID
Rn.93045
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031785.1 NP_113973.1 7, 8 915
GenBank mRNA AF035387.1 - 7, 8 915
BC070937.1 - 7, 8 915 7099117

View Full Product Details