Gene Transcripts

Gene Symbol
Fcnb
Entrez Gene ID
114091
Gene Name
ficolin B
Gene Aliases
Fcn2
Chromosome Location
Chr. 3: 6617816 - 6626193 on Build Rnor_6.0
UniGene ID
Rn.48281
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053634.1 NP_446086.1 9 919
GenBank mRNA AB036792.1 - 9 919
BC088123.1 - 9 911 7376834
FQ220557.1 - 9 922
FQ233041.1 - 9 922

View Full Product Details